ENA447955805 Enamine

1-[5-(2-methylpropyl)-1,3,4-oxadiazol-2-yl]methanamine; oxalic acid

  • MDL number MFCD28383821
  • Cas Number : 1803561-29-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HOOCCOOH
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204590
Availability details for
Quantity
  • Availability
  • Add to Cart