ENA447955806 Enamine

methyl 3-amino-2-benzylbutanoate hydrochloride

  • MDL number MFCD28383822
  • Cas Number : 1803591-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204593
Availability details for
Quantity
  • Availability
  • Add to Cart