ENA447955810 Enamine

4-(2,2-difluoroethyl)-3,4-dihydro-2H-1,4-benzothiazin-6-amine hydrochloride

  • MDL number MFCD28383828
  • Cas Number : 1803587-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204611
Availability details for
Quantity
  • Availability
  • Add to Cart