ENA447955813 Enamine

{[4-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}dimethylamine hydrochloride

  • MDL number MFCD28397398

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204642
Availability details for
Quantity
  • Availability
  • Add to Cart