ENA447955816 Enamine

3'-imino-3,4-dihydro-2H-spiro[naphthalene-1,2'-[1,4]oxazolidin]-5'-one

  • MDL number MFCD28383833
  • Cas Number : 1691964-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204662

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204662
Availability details for
Quantity
  • Availability
  • Add to Cart