ENA447955817 Enamine

methyl 4-sulfamoyl-2-(trifluoromethyl)benzoate

  • MDL number MFCD28383834
  • Cas Number : 1803561-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204679
Availability details for
Quantity
  • Availability
  • Add to Cart