ENA447955818 Enamine

1-[4-(3,5-dimethoxyphenoxy)phenyl]methanamine hydrochloride

  • MDL number MFCD28383835
  • Cas Number : 1803612-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204715
Availability details for
Quantity
  • Availability
  • Add to Cart