ENA447955819 Enamine

3-(5-fluoro-1H-indol-3-yl)propan-1-amine hydrochloride

  • MDL number MFCD28383836
  • Cas Number : 1803598-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204722
Availability details for
Quantity
  • Availability
  • Add to Cart