ENA447955825 Enamine

4-(4-chlorophenyl)-5-methyl-1H-1,2,3-triazole

  • MDL number MFCD28383843
  • Cas Number : 1803587-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204774
Availability details for
Quantity
  • Availability
  • Add to Cart