ENA447955826 Enamine

N-[(thiophen-2-yl)methyl]-2,3-dihydro-1H-inden-2-amine hydrochloride

  • MDL number MFCD28383845
  • Cas Number : 133276-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204779

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204779
Availability details for
Quantity
  • Availability
  • Add to Cart