ENA447955828 Enamine

4-(benzyloxy)pyridine-3-carboxamide

  • MDL number MFCD28383847
  • Cas Number : 100738-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204788
Availability details for
Quantity
  • Availability
  • Add to Cart