ENA447955833 Enamine

2-(4-fluoropiperidin-4-yl)pyridine dihydrochloride

  • MDL number MFCD28383853
  • Cas Number : 1803604-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204839

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204839
Availability details for
Quantity
  • Availability
  • Add to Cart