ENA447955835 Enamine

dimethyl[2-(4-nitrophenyl)ethyl]amine hydrochloride

  • MDL number MFCD28383855
  • Cas Number : 1803605-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204850
Availability details for
Quantity
  • Availability
  • Add to Cart