ENA447955839 Enamine

tert-butyl N-{2-[(2-phenylethyl)amino]ethyl}carbamate hydrochloride

  • MDL number MFCD28383859
  • Cas Number : 1803587-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204862
Availability details for
Quantity
  • Availability
  • Add to Cart