ENA447955842 Enamine

1-cyclopentyl-1-[3-(trifluoromethyl)phenyl]methanamine hydrochloride

  • MDL number MFCD28383861
  • Cas Number : 1803598-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204904
Availability details for
Quantity
  • Availability
  • Add to Cart