ENA447955843 Enamine

5-(piperidine-4-carbonyl)-2,3-dihydro-1H-indol-2-one hydrochloride

  • MDL number MFCD28383862
  • Cas Number : 1803611-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204906
Availability details for
Quantity
  • Availability
  • Add to Cart