ENA447955844 Enamine

3-(1-phenyl-1H-1,2,4-triazol-5-yl)morpholine hydrochloride

  • MDL number MFCD28383864
  • Cas Number : 1803587-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204909

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204909
Availability details for
Quantity
  • Availability
  • Add to Cart