ENA447955846 Enamine

2-[3-(4-chlorophenyl)-1H-1,2,4-triazol-5-yl]ethan-1-amine dihydrochloride

  • MDL number MFCD28383866
  • Cas Number : 1803561-96-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204911
Availability details for
Quantity
  • Availability
  • Add to Cart