ENA447955851 Enamine

[2-(1H-imidazol-1-yl)ethyl][(1-methyl-1H-pyrazol-4-yl)methyl]amine trihydrochloride

  • MDL number MFCD28383871
  • Cas Number : 1803605-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204946
Availability details for
Quantity
  • Availability
  • Add to Cart