ENA447955852 Enamine

3-(bromomethyl)quinoline hydrobromide

  • MDL number MFCD28383872
  • Cas Number : 1192352-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204951

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204951
Availability details for
Quantity
  • Availability
  • Add to Cart