ENA447955853 Enamine

3-(4-fluoropiperidin-4-yl)pyridine dihydrochloride

  • MDL number MFCD28383873
  • Cas Number : 1803587-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204952
Availability details for
Quantity
  • Availability
  • Add to Cart