ENA447955854 Enamine

5-(aminomethyl)-N-(propan-2-yl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxamide dihydrochloride

  • MDL number MFCD28383875
  • Cas Number : 1803605-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204962
Availability details for
Quantity
  • Availability
  • Add to Cart