ENA447955855 Enamine

1-[3-(4-methylpiperazine-1-carbonyl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyridin-5-yl]methanamine trihydrochloride

  • MDL number MFCD28383876
  • Cas Number : 1803592-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 3HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204967

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204967
Availability details for
Quantity
  • Availability
  • Add to Cart