ENA447955857 Enamine

2-[5-(2-fluorophenyl)-1,3,4-oxadiazol-2-yl]piperidine hydrochloride

  • MDL number MFCD28383878
  • Cas Number : 1803562-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204978
Availability details for
Quantity
  • Availability
  • Add to Cart