ENA447955859 Enamine

4-(chloromethyl)-1H-1,2,3-triazole hydrochloride

  • MDL number MFCD28383879
  • Cas Number : 1803601-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205016
Availability details for
Quantity
  • Availability
  • Add to Cart