ENA447955878 Enamine

2-oxo-2,3-dihydro-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD19219043
  • Cas Number : 857982-30-0
  • Molecular Weight : 145.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-181711

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-181711
Availability details for
Quantity
  • Availability
  • Add to Cart