ENA447955893 Enamine

2-(bromomethyl)-1,1-dichlorocyclopropane

  • MDL number MFCD00052448
  • Cas Number : 3591-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18815
Availability details for
Quantity
  • Availability
  • Add to Cart