ENA447955915 Enamine

7-bromo-2,3-dihydro-1H-isoindol-1-one

  • MDL number MFCD11848600
  • Cas Number : 200049-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191719
Availability details for
Quantity
  • Availability
  • Add to Cart