ENA447956076 Enamine

1-(methoxymethoxy)-4-(trifluoromethyl)benzene

  • MDL number MFCD19688630
  • Cas Number : 174265-24-8
  • Molecular Weight : 206.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-200565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-200565
Availability details for
Quantity
  • Availability
  • Add to Cart