ENA447956120 Enamine

2,2-dicyclobutylacetonitrile

  • MDL number MFCD28397376
  • Cas Number : 1783349-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201905
Availability details for
Quantity
  • Availability
  • Add to Cart