ENA447956125 Enamine

5-tert-butylpyridin-2-ol

  • MDL number MFCD12033221
  • Molecular Weight : 151.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-129208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-129208
Availability details for
Quantity
  • Availability
  • Add to Cart