ENA447956130 Enamine

6-methylimidazo[1,2-a]pyridine-2-carbaldehyde

  • MDL number MFCD05863353
  • Cas Number : 202348-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201958
Availability details for
Quantity
  • Availability
  • Add to Cart