ENA447956142 Enamine

4-amino-1-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}butan-1-one hydrochloride

  • MDL number MFCD28397379
  • Cas Number : 1579809-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202151
Availability details for
Quantity
  • Availability
  • Add to Cart