ENA447956144 Enamine

2-[(carboxymethyl)sulfanyl]-2-methylpropanoic acid

  • MDL number MFCD19259553
  • Cas Number : 13330-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202195
Availability details for
Quantity
  • Availability
  • Add to Cart