ENA447956153 Enamine

3,3-dimethylbutyl methanesulfonate

  • MDL number MFCD25966266
  • Cas Number : 69436-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202421
Availability details for
Quantity
  • Availability
  • Add to Cart