ENA447956162 Enamine

7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylic acid

  • MDL number MFCD26935663
  • Cas Number : 417721-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202647

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202647
Availability details for
Quantity
  • Availability
  • Add to Cart