ENA447956163 Enamine

methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate

  • MDL number MFCD13192255
  • Cas Number : 205448-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202648
Availability details for
Quantity
  • Availability
  • Add to Cart