ENA447956169 Enamine

2-[4-(2-methylbutan-2-yl)cyclohexyl]ethan-1-ol

  • MDL number MFCD28383767
  • Cas Number : 1803610-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-202763

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202763
Availability details for
Quantity
  • Availability
  • Add to Cart