ENA447956179 Enamine

5-bromo-3-methyl-1,2-thiazole-4-carbonitrile

  • MDL number MFCD27991839
  • Cas Number : 1803561-46-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203000
Availability details for
Quantity
  • Availability
  • Add to Cart