ENA447956183 Enamine

2,2,2-trichloro-1-(1H-indol-3-yl)ethan-1-amine

  • MDL number MFCD28383770
  • Cas Number : 475277-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203039
Availability details for
Quantity
  • Availability
  • Add to Cart