ENA447956189 Enamine

N,2-dihydroxy-4-methoxybenzamide

  • MDL number MFCD09942782
  • Cas Number : 90222-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 198-199
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203108
Availability details for
Quantity
  • Availability
  • Add to Cart