ENA447956194 Enamine

methyl 3-chloro-2-(chlorosulfonyl)benzoate

  • MDL number MFCD28383771
  • Cas Number : 99943-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203143
Availability details for
Quantity
  • Availability
  • Add to Cart