ENA447956197 Enamine

N,N-diethyl-3,5-dimethoxybenzamide

  • MDL number MFCD01215858
  • Cas Number : 38228-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203223
Availability details for
Quantity
  • Availability
  • Add to Cart