ENA447956201 Enamine

2,4-dichloro-6-(pyridin-2-yl)pyrimidine

  • MDL number MFCD09750202
  • Cas Number : 954232-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203323
Availability details for
Quantity
  • Availability
  • Add to Cart