ENA447956204 Enamine

2-cyclopropyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD28383774
  • Cas Number : 1484637-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203347
Availability details for
Quantity
  • Availability
  • Add to Cart