ENA447956207 Enamine

5-(dimethoxymethyl)-2-fluorobenzaldehyde

  • MDL number MFCD24678032
  • Cas Number : 334019-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203353
Availability details for
Quantity
  • Availability
  • Add to Cart