ENA447956210 Enamine

2-[(5-methyl-1,3-oxazol-2-yl)methoxy]acetic acid

  • MDL number MFCD12403816
  • Cas Number : 1216071-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203449
Availability details for
Quantity
  • Availability
  • Add to Cart