ENA447956211 Enamine

7-bromo-1,3-benzoxazole

  • MDL number MFCD08234619
  • Cas Number : 885270-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-43
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203450

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203450
Availability details for
Quantity
  • Availability
  • Add to Cart