ENA447956214 Enamine

2-{[(3-cyanophenyl)methyl](methyl)amino}acetic acid

  • MDL number MFCD12171144
  • Cas Number : 1153239-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203521

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203521
Availability details for
Quantity
  • Availability
  • Add to Cart