ENA447956223 Enamine

6-amino-5-fluoro-1,2,3,4-tetrahydroisoquinolin-1-one

  • MDL number MFCD20702794
  • Cas Number : 912846-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203614
Availability details for
Quantity
  • Availability
  • Add to Cart